Merck
Stericup® Vacuum Filters
Stericup sterile filtration devices filter media and buffers for cell culture
Stericup® filtration devices have been the most trusted systems in media and buffer filtration for decades. The next-generation Stericup® Quick Release vacuum filtration devices present ergonomic design updates that optimize usability and streamline the filtration process, while protecting essential cell cultures with the renowned performance of Millipore® membranes.   
The minds of Millipore® scientists don’t stop with the most reliable sterile filtration in the world. Our commitment to sustainability drove us to design a dependable sterile filter device that helps labs attain sustainability goals. Stericup® E and Steritop® E filters provide the same steadfast sterile filtration cell culturists worldwide have chosen for decades—but attach directly to commercial media bottles, eliminating the plastic filter funnel to reduce plastic waste up to 48%.
View our product offering or request information below.
POPULAR CATEGORIES
Products
Compare
Product Number
Product Name
Product Description
Pricing
Related Product Resources
Page 1 of 2
Stericup® &  Steritop® Filter Units
Stericup® and Steritop® sterile filtration devices use reliable, high-performance membranes for optimum sterile filtration of laboratory reagents, buffers, and media. These devices combine superior flow rates and throughput with low non-specific binding and a stable, no-tip design.
  • Stericup® Filter Units combine a filter unit with a receiver flask and cap for processing and storage
  • Steritop® Bottle-top Filter Units can be used on bottles with 33 mm or 45 mm openings
  • Click Seal Receiver Bottles and Caps are available in 250 mL, 500 mL, 1000 mL Receiver Bottles
Work with ease and filter with confidence with Stericup® and Steritop® Filtration Systems.  View all products.
Applications
Stericup® and Steritop® systems are designed to reduce foaming and protein denaturation while maximizing flow and can be used for sterile vacuum filtration of aqueous solutions such as tissue culture media and biological fluids.
  • Tissue culture media +/- additives
  • Buffers
  • Biological solutions
Fit-for-Application Membrane Chemistries
Sterile filtration performance depends on the quality of the filter membrane. Stericup® and Steritop® filters are available with a selection of Millipore® membranes that set the industry standard for their application-specific properties.
  • Fastest flow, low protein-binding of aqueous solutions with Millipore Express® and Express® Plus polyethersulfone (PES) membrane devices
  • Broad chemical compatibility and very low protein binding polyvinylidene fluoride (PVDF)
Membrane Pore Size
Pore size relates to the filter’s ability to filter out particles of a certain size. Stericup® and Steritop® filters can be selected by pore size to meet your specific needs.
  • 0.1 µm: mycoplasma elimination
  • 0.22 µm: sterilizing filtration
  • 0.45 µm: clarification
Process and Storage Volumes
Process volumes are based on funnel capacity for Steritop® filter units or a combination of funnel capacity and matching receiver bottle capacity for Stericup® filter units. Stericup® and Steritop® filtration devices can be used to process and store volumes ranging from 150 mL to 1 L.
  • 150 mL
  • 250 mL
  • 500 mL
  • 1000 mL
Break down of the design of the Stericup® Quick Release Filtration System into 4 parts.
Figure 1.Stericup® Quick Release Filtration System.
Stericup® Quick Release Filtration Systems Design
  1. The Quick Release filter funnel disconnects from the receiver bottle with just a quarter turn—reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood.
  2. Bold product identification on the Steritop® filter funnel plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection.
  3. Click seal cap is flanged and modified to enable a firm grip in wet or dry conditions—and a tactile stop confirms secure closure. Worry less about the possibility of spilling or contamination of sterilized contents.
  4. Frosted, ample writing surface on the receiver bottle and lighter cap color facilitate clear labelling to improve legibility and reduce unreadable smears.
Additional Convenience
  • Click seal cap rests on the side to avoid risk of contamination.
  • Receiver bottles are stackable to save storage space.
Stericup® filters provide fast flow rate with low protein retention
Filtration performance depends on membrane performance. Our Millipore Express® PLUS, Durapore®, MF-Millipore™ and Fluoropore™ brand membranes set the industry standard for quality. Combined with the ergonomic improvements of the Quick Release design, Stericup® and Steritop® sterile filtration systems are ideal for your filtration needs. View this video to learn more about the quality features of our Quick Release filtration devices.
Stericup filters filter fast, with low protein
Figure 1.Stericup® filters provide fast flow rate with low protein retention
Stericup filters filter fast, with low protein
Figure 1.Stericup® filters provide fast flow rate with low protein retention
Stericup® E and Steritop® E devices are a greener choice for dependable sterile filtration
Stericup® and Steritop® E Filters Use Less Plastic and Packaging
Our engineers found a device design that retains exceptional Stericup® filtration while providing your lab a way to dramatically reduce the use of disposable plastic and packaging materials. Whether your goal is a green lab certification, or just a smaller footprint, both the Stericup® E and Steritop® E filters are designed to ensure trouble-free cell culture, while diminishing environmental impact in multiple ways.
Stericup® E devices flip sterile filtration on its head—literally. We’ve eliminated the need for the plastic filler funnel by creating a way to attach standard media bottles or glassware directly onto the filter unit. Simply invert the Stericup® E filter/receiver bottle assembly and thread onto the bottle containing media or buffer.  Flip the assembly over, attach to vacuum, and watch sterile filtration happen. The Steritop® E device saves even more plastic by allowing you to attach the filter to your own compatible, reusable labware receiver bottle before threading onto the bottle containing unfiltered media.
Learn More
Stericup® E and Steritop® E filters provide significant waste reduction to help labs achieve sustainability goals:
  • Up to 26% reduction in plastic waste for Stericup® E devices, and up to 48% plastic reduction for Steritop® E filters when compared with traditional plastic filters 
  • Because devices are smaller, packaging is reduced by up to 48% for Stericup® E filters, and by 69% for Steritop® E devices