Skip to Content
Merck
All Photos(1)

Key Documents

PHL83478

Aristolochic acid D

phyproof® Reference Substance

Synonym(s):

10-Hydroxy-8-methoxy-6-nitrophenanthro[3,4-d]-1,3-dioxole-5-carboxylic acid, Aristolochic acid Iva

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11NO8
CAS Number:
Molecular Weight:
357.27
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

2-8°C

InChI

1S/C17H11NO8/c1-24-12-3-7(19)2-9-8(12)4-11(18(22)23)14-10(17(20)21)5-13-16(15(9)14)26-6-25-13/h2-5,19H,6H2,1H3,(H,20,21)

InChI key

PADIFGYTAXNCRK-UHFFFAOYSA-N

General description

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service