Skip to Content
Merck
All Photos(1)

Key Documents

SCP0236

Sigma-Aldrich

Renin Substrate I

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H87N14O14
Molecular Weight:
1300.48
UNSPSC Code:
12352204
NACRES:
NA.32

product name

Renin Substrate I,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Suc-Arg-Pro-Phe-His-Leu-Leu-Val-Tyr-AMC

Application

Renin (angiotensinogenase) is a component of the renin-angiotensin system (RAS) that regulates vasoconstriction and blood pressure. Renin which hydrolyzes angiotensinogen in vivo may be detected using peptides substrates such as succinyl-RPFHLLVY-AMC. This substrate may also be used to detect proteinase A from yeast.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service