Skip to Content
Merck
All Photos(1)

Key Documents

R9153

Sigma-Aldrich

Anti-Rhodopsin antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1-3 μg/mL (paraffin-embedded sections)

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RHO(6010)
mouse ... Rho(212541)
rat ... Rho(24717)

Immunogen

synthetic peptide corresponding to the second extracellular loop of human rhodopsin, conjugated to KLH. The immunizing peptide has 100% homology with the rat and mouse gene.

Physical form

Solution in phosphate buffered saline containing 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caroline J Zeiss et al.
Journal of morphology, 272(8), 949-957 (2011-05-14)
The purpose of this study is to identify evolutionary origin and fate of anatomic features of the duck-billed platypus eye. Eyes from the duck-billed platypus and four key evolutionary basal vertebrates (Pacific hagfish, north hemisphere sea lamprey, and Australian and
Charles S Schobert et al.
Veterinary ophthalmology, 16(5), 359-364 (2012-11-29)
Troglomorphic fishes provide excellent comparative models for studying eye evolution. We describe the gross and microscopic anatomy of ocular structures of the depigmented, blind cichlid, Lamprologus lethops, and its putative sister species, Lamprologus tigripictilis collected from the lower Congo River

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service