Skip to Content
Merck
All Photos(1)

Key Documents

GE27-4110-02

Poly (A)

Cytiva 27-4110-02, pack of 500 mg

Synonym(s):

Poly(A) RNA, Polyadenylic acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006
NACRES:
NA.51

packaging

pack of 500 mg

manufacturer/tradename

Cytiva 27-4110-02

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

Poly(A) is a ribonuclease tested, single-stranded RNA homopolymer, with an average chain length of >200 nucleotides.

Biochem/physiol Actions

In drug development research, poly(A) (polyadenylic acid) can be employed to assess the binding of potential therapeutic drugs to mRNA.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service