ERMBC516
Apple (dietary fibre)
ERM®, certified reference material
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
certified reference material
Agency
ERM®
certified by the BSC
manufacturer/tradename
JRC
application(s)
food and beverages
format
matrix material
storage temp.
−20°C
Analysis Note
For more information please see:
ERMBC516
ERMBC516
Other Notes
Certified for the analytes listed below. See certificate for values and more details
Dietary Fibre: Dietary fibre according to AOAC 1990985.29, Dietary fibre according to AOAC 1992 MES-TRIS 991.43, Dietary fibre according to Englyst (by Colorimetry), Dietary fibre according to Englyst (by GC), Dietary fibre according to Uppsala 994.13
Matrix Group: Fruits and Vegetables
Dietary Fibre: Dietary fibre according to AOAC 1990985.29, Dietary fibre according to AOAC 1992 MES-TRIS 991.43, Dietary fibre according to Englyst (by Colorimetry), Dietary fibre according to Englyst (by GC), Dietary fibre according to Uppsala 994.13
Matrix Group: Fruits and Vegetables
Legal Information
ERM is a registered trademark of European Commission
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service