48915-U
ProClin™ 300
ProClin™ preservative for use in IVD manufacturing, pkg of 2.0 L, CMIT/MIT 3 %
Synonym(s):
3% solution of 5-chloro-2-methyl-4-isothiazolin-3-one and 2-Methyl-4-isothiazolin-3-one
About This Item
Quality Level
form
liquid
shelf life
3 yr
contains
Alkyl Carboxylate
composition
CMIT/MIT, 3%
packaging
pkg of 2.0 L
parameter
<45 °C operating temperature
color
colorless to light yellow
matrix
Modified glycol
operating pH range
2.5-8.5
pH
4.1 (100 g/L)
bp
189 °C
mp
-40 °C
solubility
water: soluble
density
1.03 g/cm3
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
244.4 °F - closed cup
Flash Point(C)
118 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service