Skip to Content
Merck
All Photos(1)

Documents

MABF607

Sigma-Aldrich

Anti-CD45R (B220) Antibody (human/mouse), violetFluor® 450, clone RA3-6B2

clone RA3-6B2, 0.2 mg/mL, violetFluor®

Synonym(s):

Receptor-type tyrosine-protein phosphatase C, Leukocyte common antigen, L-CA, Lymphocyte antigen 5, Ly-5, T200, CD45

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

conjugate

violetFluor® 450

antibody form

purified antibody

antibody product type

primary antibodies

clone

RA3-6B2, monoclonal

species reactivity

human, mouse

manufacturer/tradename

violetFluor®

concentration

0.2 mg/mL

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PTPRC(5788)
mouse ... Ptprc(19264)

Related Categories

General description

The RA3-6B2 antibody reacts with the human and mouse CD45 isoform known as CD45R, or B220, a protein tyrosine phosphatase of 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns, all critical for leukocyte function. In mouse, the CD45R/B220 isoform is predominantly found on B cells, at varying levels on all stages from pro-B cells to activated B cells, and may also be detected on certain T cell and NK cell subsets. It is of note that B220 is not similarly expressed on human B cells, where it appears to be differentiation-specific and therefore expressed on only some B cell subsets. Other forms of CD45 with restricted cellular expression include CD45RA, CD45RB, CD45RO and several others.The RA3-6B2 antibody is one of the most consistently used leukocyte markers for B cells, T cell subsets and NK cell subsets in human and mouse. This antibody has also been reported as cross-reactive with feline CD45R/B220.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
This rat monoclonal antibody Anti-CD45R (B220) (human/mouse), violetFluor 450, clone RA3-6B2 validated for use in western blotting, IP, IHC, Immunofluorescence & Flow Cytometry for the detection of CD45R (B220).

Quality

The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.5 µg of this antibody detected CD45R (B220) in one million C57Bl/6 splenocytes.

Physical form

Purified Rat Monoclonal Antibody in buffer containing 10 mM NaH2PO4, 150 mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Size Exclusion Chromatography

Storage and Stability

It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Legal Information

violetFluor is a registered trademark of Tonbo Biotechnologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Wen Lo et al.
Frontiers in immunology, 12, 636818-636818 (2021-05-28)
In addition to regulating immune responses by producing antibodies that confer humoral immunity, B cells can also affect these responses by producing cytokines. How B cells participate in the clearance of pathogenic infections via functions other than the production of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service