Merck
HPLC Analysis of 25-Hydroxyvitamin D3 and epi-25-Hydroxyvitamin D3 on Ascentis® Express F5
HPLC Analysis of 25-Hydroxyvitamin D3 and epi-25-Hydroxyvitamin D3 on Ascentis® Express F5 application for HPLC
材料
分析色谱柱
产品编号
说明
价格
Ascentis® Express F5, 2.7 μm HPLC 色谱柱
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
标准
产品编号
说明
价格
3--25-羟基维生素 D3
98% (CP)
25-羟基维生素D3标准液 CRM 溶液
100 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
流动相组分
产品编号
说明
价格
甲酸铵
eluent additive for LC-MS, LiChropur, ≥99.0%
CONDITIONS
column
Ascentis Express F5, 10 cm x 2.1 mm I.D., 2.7 μm particles (53569-U)
mobile phase
[A] 7 mM ammonium formate; [B] methanol; (25:75, A:B)
flow rate
0.4 mL/min
column temp.
40 °C
detector
UV, 265 nm
ESI(+), m/z 100 - 1000
injection
1 μL
sample
300 mg/L in 25:75, water:methanol