Merck
Pestizide in Obst und Gemüse
Pestizide in Obst und Gemüse suitable for GC, application for SPE
材料
SPE管或板
产品编号
说明
价格
Supelclean ENVI-Carb SPE 管
bed wt. 500 mg, volume 6 mL, pk of 30
CONDITIONS
sample preparation
SPE (Solid Phase Extraction)
sample/matrix
homogenize 50 g sample in 100 mL acetonitrile then add 10 g NaCl and homogenize 5 min
SPE tube/cartridge
ENVI-Carb, 500 mg/6 mL
column
14% cyanopropylphenyl polysiloxane, 30 m × 0.25 mm I.D., 0.15 μm (Supelco equivalent, Equity-1701, 28372-U)
oven
70 °C (2 min), 25 °C/min to 130 °C, 2 °C/min to 220 °C, 10 °C/min to 280 °C (4.6 min)
detector
MSD, 285 °C
carrier gas
helium
injection
2 μL splitless