跳转至内容
Merck
  • SIRT3 blocks myofibroblast differentiation and pulmonary fibrosis by preventing mitochondrial DNA damage.
SIRT3 blocks myofibroblast differentiation and pulmonary fibrosis by preventing mitochondrial DNA damage.
American journal of physiology. Lung cellular and molecular physiology (2016-11-07)
Samik Bindu, Vinodkumar B Pillai, Abhinav Kanwal, Sadhana Samant, Gökhan M Mutlu, Eric Verdin, Nickolai Dulin, Mahesh P Gupta
摘要
Myofibroblast differentiation is a key process in the pathogenesis of fibrotic diseases. Transforming growth factor-β
材料
货号
品牌
产品描述
Sigma-Aldrich
MISSION® esiRNA, targeting human OGG1