跳转至内容
Merck
Coffee silverskin: a possible valuable cosmetic ingredient.
Pharmaceutical biology (2014-12-05)
Francisca Rodrigues, Ana Palmeira-de-Oliveira, José das Neves, Bruno Sarmento, M Helena Amaral, M Beatriz P P Oliveira
摘要
Currently, there is a great tendency in cosmetic area to use natural extracts. Coffee silverskin (CS) is the most abundant solid by-product generated during roasting of coffee processing. To evaluate different CS extracts as promising cosmetic ingredients, regarding antioxidant, antimicrobial, and cytotoxic properties. Aqueous, hydroalcoholic and ethanolic CS extracts were obtained by an environmentally friendly procedure considering costs and pollution. Extracts were characterized for total phenolic and flavonoid contents (TPC and TFC, respectively), antioxidant activity by 1,1-diphenyl-2-picrylhydrazyl (DPPH), ferric reducing antioxidant power (FRAP), antimicrobial activity expressed as minimal inhibitory concentration (MIC) and cytotoxicity using the 3-(4,5-dimethylthiazol-2-yl)-5-(3-carboxymethoxyphenyl)-2-(4-sulfophenyl)-2H-tetrazolium (MTS) and lactate dehydrogenase (LDH) assays in two skin cell lines (fibroblasts and keratinocytes). The TPC of extracts was 18.33-35.25 mg of gallic acid equivalents per g of material on a dry basis (mg GAE/g db). The TFC of extracts was 1.08-2.47 µg cathechin equivalents per g dry material (µg CE/g db). The antioxidant activity was high, with values ranging between 95.95 and 216.40 µmol Fe(2+)/g for aqueous and alcoholic samples, respectively. Preliminary assays for antimicrobial potential showed that extracts display antibacterial activity. The MIC varied from 31.3 to 250 µg/mL for Gram-positive, and from 31.3 to 1000 µg/mL for Gram-negative. Extracts did not affect in vitro cell viability, with values near 100% in all concentrations tested. RESULTS seem show that CS is a safe source of natural antioxidants with antifungal and antibacterial activity and no cytotoxicity, with potential usefulness for cosmetic applications.
材料
货号
品牌
产品描述
Sigma-Aldrich
纯乙醇, for molecular biology
Sigma-Aldrich
纯乙醇, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
甘油, ACS reagent, ≥99.5%
Sigma-Aldrich
氢氧化钠, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
甘油, for molecular biology, ≥99.0%
Sigma-Aldrich
纯乙醇, 200 proof
Sigma-Aldrich
氢氧化钠, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
甘油, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
氢氧化钠 溶液, 50% in H2O
Sigma-Aldrich
碘, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
无水碳酸钠, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
D -(+)-葡萄糖, ≥99.5% (GC)
Sigma-Aldrich
纯乙醇, 200 proof, meets USP testing specifications
Sigma-Aldrich
乙酸钠, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
D -(+)-葡萄糖, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
无水碳酸钠, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
2,2-联苯基-1-苦基肼基
Sigma-Aldrich
亚硝酸钠, ReagentPlus®, ≥99.0%
Sigma-Aldrich
亚硝酸钠, ACS reagent, ≥97.0%
Sigma-Aldrich
氢氧化钠 溶液, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
纯乙醇, for molecular biology
Sigma-Aldrich
乙酸钠, ACS reagent, ≥99.0%
Sigma-Aldrich
乙酸钠, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
L -抗坏血酸, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L -抗坏血酸, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
氢氧化钠, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
氯化铝, ReagentPlus®, 99%
Sigma-Aldrich
氢氧化钠, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
氢氧化钠 溶液, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
葡萄糖, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications