跳转至内容
Merck
  • Improvement of Salmonella detection on motility enrichment media by ferrioxamine E-supplementation of pre-enrichment culture.
Improvement of Salmonella detection on motility enrichment media by ferrioxamine E-supplementation of pre-enrichment culture.
International journal of food microbiology (1995-10-01)
P Pless, R Reissbrodt
摘要
Supplementation of buffered peptone water with ferrioxamine E in concentrations ranging from 0.001 to 1.0 microgram/ml significantly increased the motility of Salmonella on MSRV and DIASSALM semi-solid enrichment media. Zone diameters of swarming S. enteritidis and S. typhimurium increased more than twofold following use of pre-enrichment cultures supplemented with 0.01 microgram ferrioxamine E/ml. The activity of ferrioxamine E is similar at 37 degrees C and 42 degrees C. Pre-enrichment of Salmonella in a variety of foods in supplemented buffered peptone water, with shaking at 37 degrees C for 6 h and motility enrichment at 42 degrees C for 16 h, enabled motile Salmonella to be detected in 1 day.
材料
货号
品牌
产品描述
Millipore
铁草氨菌素 E 来源于抗生素链霉菌, suitable for microbiology, ≥95%