跳转至内容
Merck
  • Eplerenone is not superior to older and less expensive aldosterone antagonists.
Eplerenone is not superior to older and less expensive aldosterone antagonists.
The American journal of medicine (2012-07-31)
Saurav Chatterjee, Chaim Moeller, Nidhi Shah, Oluwaseyi Bolorunduro, Edgar Lichstein, Norbert Moskovits, Debabrata Mukherjee
摘要
Eplerenone is publicized to be extremely effective in reducing mortality from heart failure, with a reasonable side-effect profile. However, it is much more expensive compared with older aldosterone antagonists. We reviewed available evidence to assess whether increased expense was justified with outcomes data. The authors searched the PubMed, CENTRAL, CINAHL, and EMBASE databases for randomized controlled trials from 1966 through July 2011. Interventions included aldosterone antagonists (Aldactone [Pfizer, NY, NY], canrenone, eplerenone) in systolic heart failure. The comparator included standard medical therapy or placebo, or both. Outcomes assessed were mortality in the intervention versus the comparator groups, and rates of adverse events at the end of at least 8 weeks of follow-up. Event rates were compared using a forest plot of relative risk (RR) (95% confidence interval [CI]) using a random-effects model (Mantel-Haenszel) between the aldosterone antagonists and controls. We included 13 studies for aldosterone antagonists other than eplerenone, and 3 studies for eplerenone. There was significant reduction of mortality with all aldosterone antagonists, but eplerenone (15% mortality relative reduction; RR 0.85; 95% CI, 0.77-0.93; P=.0007) was outperformed by other aldosterone antagonists, namely, spironolactone and canrenone (26% mortality relative reduction; RR 0.74; 95% CI, 0.66-0.83; P <.0001). Reduction in cardiovascular mortality with eplerenone was 17% (RR 0.83; 95% CI, 0.75-0.92; P=.0005), while that with other aldosterone antagonists was 25% (RR 0.75; 95% CI, 0.67-0.84, P <.0001), without contributing significantly to an improved side-effect profile. Eplerenone does not appear to be more effective in reducing clinical events compared with older, less expensive aldosterone antagonists.
材料
货号
品牌
产品描述
Canrenone, European Pharmacopoeia (EP) Reference Standard