跳转至内容
Merck
Ultralarge single crystal SnS rectangular nanosheets.
Chemical communications (Cambridge, England) (2011-03-23)
Yejun Zhang, Jun Lu, Shuling Shen, Huarui Xu, Qiangbin Wang
摘要
Uniform ultralarge single crystal SnS rectangular nanosheets (7000 nm × 3000 nm × 20 nm) have been synthesized by thermodecomposing a single-source precursor (Sn-diethyldithiocarbamate-1,10-phenanthroline). The obtained SnS nanosheets exhibit excellent electrochemical properties which have promising applications in lithium ion batteries.