跳转至内容
Merck
JMJD6 is a histone arginine demethylase.
Science (New York, N.Y.) (2007-10-20)
Bingsheng Chang, Yue Chen, Yingming Zhao, Richard K Bruick
摘要
Arginine methylation occurs on a number of proteins involved in a variety of cellular functions. Histone tails are known to be mono- and dimethylated on multiple arginine residues where they influence chromatin remodeling and gene expression. To date, no enzyme has been shown to reverse these regulatory modifications. We demonstrate that the Jumonji domain-containing 6 protein (JMJD6) is a JmjC-containing iron- and 2-oxoglutarate-dependent dioxygenase that demethylates histone H3 at arginine 2 (H3R2) and histone H4 at arginine 3 (H4R3) in both biochemical and cell-based assays. These findings may help explain the many developmental defects observed in the JMJD6(-/-) knockout mice.
材料
货号
品牌
产品描述
Sigma-Aldrich
JMJD6 human, recombinant, expressed in baculovirus infected Sf9 cells, ≥80% (SDS-PAGE)