跳转至内容
Merck
  • Synthesis and Characterization of Thiolated PVP-Iodine Complexes: Key to Highly Mucoadhesive Antimicrobial Gels.
Synthesis and Characterization of Thiolated PVP-Iodine Complexes: Key to Highly Mucoadhesive Antimicrobial Gels.
Molecular pharmaceutics (2018-07-27)
Aamir Jalil, Barbara Matuszczak, Nguyet-Minh Nguyen Le, Arshad Mahmood, Flavia Laffleur, Andreas Bernkop-Schnürch
摘要
The aim of this study was to synthesize iodine containing polymeric excipients for mucosal treatment of microbial infection exhibiting a prolonged mucosal residence time by forming an adhesive gel on the mucosal surface. In order to achieve this aim, 2-(2 acryloylamino-ethyldisulfanyl)-nicotinic acid (ACENA) was copolymerized with N-vinylpyrrolidone (NVP) to obtain thiolated polyvinylpyrrolidone (PVP) for complexation with iodine. The average molecular mass of different thiolated PVP variants was determined by size exclusion chromatography. The structure of thiolated PVP was confirmed by 1H NMR. Thiolated PVP variants were characterized for thiol content, cytotoxicity, iodine loading capacity, rheological behavior, and adhesion time on mucosa. The highest achieved degree of thiolation was 610 ± 43 μmol/g, and the maximum recorded iodine loading was 949 ± 31 μmol/g of polymer. Thiolated PVP variants (0.5% m/v) showed no toxicity after incubation on Caco-2 cells for the period of 3 and 24 h, respectively. Thiolated PVP and thiolated PVP-iodine complexes exhibited a 5.4- and 4.4-fold increased dynamic viscosity in porcine mucus in comparison to PVP and PVP-iodine complex, respectively. Compared to PVP and PVP-iodine complex thiol-functionalized PVP and PVP-iodine complexes demonstrated significantly prolonged attachment to mucosal surface over a period of 3 h. Thiol functionalized PVP proved to be a promising novel excipient for complexation with iodine and to exhibit strongly improved mucoadhesive properties.
材料
货号
品牌
产品描述
Sigma-Aldrich
2-巯基-3-吡啶甲酸, technical grade