主要文件
EP5810763002
Eppendorf® Adapter for 250 mL Rectangular Bucket
for use with 4x50ml Falcon-tubes (without aerosol- tight caps), set of 2 pieces
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103911
NACRES:
NB.82
特点
for use with 4x50ml Falcon-tubes (without aerosol- tight caps)
包装
set of 2 pieces
制造商/商品名称
Eppendorf® 5810763002
技术
centrifugation: suitable
相容性
for use with 250 mL rectangular buckets
正在寻找类似产品? 访问 产品对比指南
一般描述
Tube adapter for Eppendorf 250 mL rectangular bucket.
法律信息
Eppendorf is a registered trademark of Eppendorf AG
历史批次信息供参考:
分析证书(COA)