跳转至内容
Merck
所有图片(1)

主要文件

26692

Sigma-Aldrich

Cholera Filtrate from Vibrio cholerae

lyophilized from 5 ml vibrio culture fluid

登录查看公司和协议定价


About This Item

MDL编号:
UNSPSC代码:
12352204

质量

lyophilized from 5 ml vibrio culture fluid

溶解性

0.2% BSA: 2 mg/mL, very faintly hazy, yellow

异质活性

neuraminidase ~0.1 U/vial

储存温度

2-8°C

单位定义

1 U corresponds to the amount of enzyme which releases 1μmol of N-acetylneuraminic acid per minute at pH 4.5 and 37°C (Neu5Acα(2-3,6)Gal(1-4)Glc as substrate)

其他说明

Used for the serological testing (research use only) for influenza after reconstitution with 5 mL sterile water; it will then contain 0.01 % thimerosal as preservative

历史批次信息供参考:

分析证书(COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.

如需帮助,请联系 客户支持

已有该产品?

在文件库中查找您最近购买产品的文档。

访问文档库

World Health Organ., WHO Expert Com. on Influenza, WHO Tech. Rep. Ser., 64-64 (1953)

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门