SP160015
1 of 10
Millipore
SP160029
Fastener/Screw Kit. Box Of Each Type Of Screw Used
Sigma-Aldrich
612080
TIMP-1, Human Neutrophil Granulocyte
DMS 53
SAFC
103773
内切核酸酶 来源于粘质沙雷氏菌
CLS431682
Corning® vent cap, for 5 L Erlenmeyer flasks
802956
2-Ethynylpyrimidine
251M-1
Factor XIIIa (AC-1A1) Mouse Monoclonal Antibody
CHGE72TS3
Millipore Express SHC 筒式过滤器 20 in. 0.5/0.2 µm 7 号 硅树脂
IT5SP0012
Fuse Glass 2A 250VAC 5×20MM
KHGES03TT3
Millipore Express® SHC,Opticap® XL囊式
SP1
Bioprocessing Systems
Filtration for Bioprocessing
Integrity Testers
Sterile Filtration and Bioburden Control
Viral Clearance