Merck
HomeGas Chromatography (GC) GC Analysis of Bacterial Acid Methyl Esters (BAMEs)on Equity®-1
GC Analysis of Bacterial Acid Methyl Esters (BAMEs)on Equity®-1
GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1
Conditions
columnEquity-1, 30 m x 0.25 mm I.D., 0.25 µm (28046-U)
oven150 °C (4 min), 4 °C/min to 250C (5 min)
carrier gashelium, 20 cm/sec @ 150 °C
sample100 ng on-column of a 26 component BAME standard (47080-U) 
injection1 µL, split 100:1
linersplit, cup design
inj. temp.250 °C
detectorFID, 280 °C
Description
CategoriesAnalytical Chromatography, GC Applications, Bacterial Acid Methyl Esters (BAMEs)
Featured IndustryClinical
Other NotesComplementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitabilityapplication for GC
Analyte
  • Methyl 12-methyltetradecanoate
  • Methyl 13-methyltetradecanoate
  • Methyl 14-methylpentadecanoate
  • Methyl 15-methylhexadecanoate
  • Methyl 2-hydroxydecanoate
  • Methyl 2-hydroxydodecanoate
  • Methyl 2-hydroxyhexadecanoate
  • Methyl 2-hydroxytetradecanoate
  • Methyl 3-hydroxydodecanoate
  • Methyl 3-hydroxytetradecanoate
  • Methyl cis-9-hexadecenoate
  • Methyl dodecanoate
  • Methyl eicosenoate
  • Methyl heptadecanoate
  • Methyl hexadecenoate
  • Methyl nonadecanoate
  • Methyl pentadecanoate
  • Methyl tetradecanoate
  • Methyl tridecanoate
  • Methyl undecanoate
  • Methyl cis-9,10-methylenehexadecanoate
  • Methyl cis-9,10-methyleneoctadecanoate
  • Methyl cis-9,12-octadecadienoate
  • Methyl cis-9-octadecanoate
  • Methyl octadecanoate
  • Methyl trans-9-octadecenoate
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch