Merck
HomeApplicationsProtein Biology(ELISA) Enzyme-Linked Immunosorbent Assay
(ELISA) Enzyme-Linked Immunosorbent Assay
Typically performed in multi-well microtiter plates, ELISAs are a molecular biology assay commonly used for the detection and quantification of diverse molecules, including peptides, proteins, and antibodies. Such assays can detect molecules of interest at the pg/mL level and are critical for both basic research and disease research application needs.
ELISA Assays: Antibody-Antigen Interactions
The fundamental molecular components of an ELISA typically include the use of antibody conjugated to an enzyme, an immobilized molecule(s) of interest, and a detection substrate. A critical aspect that determines the success and quality of data obtained from an ELISA is dependent on the affinity and specificity of antibody-antigen interactions. Antigen-antibody interactions are influenced by numerous factors, including pH, temperature, and ionic strength.
Something went wrong, please try again.
Featured Categories
MILLIPLEX® Multiplex Immunoassays
MILLIPLEX® Multiplex Immunoassays for Luminex® Instrumentation
Discover with confidence using MILLIPLEX® multiplex immunoassays for Luminex® instruments while ensuring robust assay performance and an always expanding portfolio to meet your biomarker detection needs.
Shop Now
ELISA Kits and Reagents
Enzyme-Linked Immunosorbent Assay (ELISA) Kits
ELISA kits, reagents, and resources for biomarker detection.
Shop Now
SMC® Immunoassays
SMC® Technology
Single Molecule Counting (SMC®) technology precisely quantifies molecules at previously undetectable levels.
Shop Now
ELISA Detection Formats
ELISAs that use direct detection methods require an immobilized antigen that is bound directly to the surface of an assay plate or indirectly by a capture antibody, followed by an antigen-specific primary antibody conjugated to an enzyme, and the detection substrate.
The more commonly used indirect detection format incorporates both an unconjugated primary antibody, followed by a conjugated secondary antibody that is specific to the detection of the primary antibody. Indirect detection benefits from increased immunoreactivity with the target antigen as the conjugated enzyme element is only present on the secondary antibody.
In addition to direct and indirect detection methods, capture or “sandwich” assays use an additional antigen-capturing antibody that is first attached to the microplate surface, followed by the use of both a primary and an enzyme-conjugated secondary antibody, similar to the indirect method previously described. 
Document Search
Looking for More Specific Information?
Visit our document search for data sheets, certificates and technical documentation.
Find Documents