Merck
HomeGene Expression & SilencingAntibiotic Optimization Using a Cytotoxicity Profile
Antibiotic Optimization for Cell Transduction Using a Cytotoxicity Profile
Cytotoxicity Profile for Optimizing Antibiotic Concentration
The appropriate concentration of antibiotic for selecting stable cell lines is different for each cell type. If the concentration for the desired cell type is unknown, a titration experiment must be performed to determine the lowest concentration of antibiotic needed to efficiently select transduced cells. In this protocol, we highlight the use of puromycin, the antibiotic used with the standard pLKO.1 vector. Typically, 1-10 µg/mL are sufficient to kill most untransduced mammalian cell types.  This general protocol can also be used for determining the optimal concentration of G418 required for the custom shRNA vectors. Higher antibiotic concentrations than required could result in off-target effects and fewer cells for downstream analysis.
Materials
  • Cells in log growth and at 50% confluence on the day of transfection
  • Cell Culture Media
  • Tissue culture incubator: 37 °C, 5% CO2, 100% relative humidity
  • Antibiotics puromycin (Product No. P9620) or G418 (Product No. A1720)
CYTOTOXICITY PROFILE PROTOCOL
Day 0
0.1Seed cells at 50% confluence per well in complete media. Seed cells in 3 rows x 11 columns for cytotoxicity assay.
0.2Return cells to incubator and incubate overnight.
Day 1
1.1Pre-warm 5 mL of full media.
1.2Make three independent dilutions in complete media of puromycin yielding 11 concentrations (µg/mL) of puromycin: 0, 1, 2, 3, 4, 5, 6 7, 8, 9, 10. Each dilution should have at least 110 µL as the final volume.
1.3Remove cells to incubator
1.4Remove media from cells
1.5Replace media with antibiotic containing media
1.6Return steps 1.1 through 1.6
Day 3
3.1Repeat steps 1.1 through 1.6.
Day 5
5.1Repeat steps 1.1 through 1.6.
Day 7
7.1Repeat steps 1.1 through 1.6.
Day 10
10.1Determine viability in each well by either cell counting or viability assay.
10.2The minimum concentration of puromycin resulting in complete cell death after 7-10 days of selection with puromycin should be used for that cell type for transduction with shRNA viral particles.
Notes:
  • If the cells started to round but did not detach from the surface after adding puromycin, the cells might be dying without detaching from the surface yet. These cells should detach with more time.
  • If cells require higher concentration of puromycin than the recommended 1-10 μg/mL, check the expiration date on the puromycin and avoid multiple freeze-thaw cycles (<5).
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch