Thorough Washing with the BioTek® 405™TSUVS Plate Washer For SMC® and MILLIPLEX® Assays
Figure 1.The BioTek® 405™TSUVS Plate Washer for SMC® and MILLIPLEX® Assays.
Ensure consistent performance of your SMC®and MILLIPLEX® assays using the BioTek® 405™TSUVS Plate Washer designed for use with SMC® technology and now for use with MILLIPLEX® assays, for thorough washing.
Section Overview
How Is the BioTek® 405™TSUVS Plate Washer Specific for SMC® Assays?
When counting at the single molecule level, it is critical to ensure reliable assay performance. For SMC® assays, one vital consideration is to ensure thorough bead washing during key stages of the workflow.
The BioTek® 405™TSUVS Magnetic 96-Well Washer has benefits specifically for SMC® technology such as:
- Customized plate washing protocols designed to optimize performance and prevent bead loss for SMC® workflows
- A custom magnetic plate uniquely manufactured for SMC® assays
- A vacuum regulator designed for use on SMC® assays
- Bottle kit
How Is the BioTek® 405™TSUVS Plate Washer Specific for MILLIPLEX® Assays?
The plate washer is easily adaptable for use in the MILLIPLEX® multiplex bead-based assay workflow based on Luminex® xMAP® technology by exchanging the custom SMC® magnetic plate on the washer with a Dexter 96F magnet (Part Number: 7103016, available from Agilent) or Dexter 384F magnet (Part Number: 7103017, available from Agilent).
Customized MILLIPLEX® wash protocols are pre-programmed on the washer for use with Luminex® assays and no physical adjustments to the washer are required.
Plate Washer Features
In addition to the unique features of the washer design, this plate washer has the same features available on the standard BioTek® 405™TSUVS Washer, a globally recognized standard for microplate washers. This includes unique features designed to wash microsphere-based assays efficiently and effectively with outstanding results for every application.
Adjustable Settings
Angled dispense tubes and adjustable dispense and aspiration rate settings to optimize washing.
Ultrasonic Cleaner
A patented ultrasonic bath enables the instrument to automatically clean buildup in the manifold preventing manifold tube clogs.
Flexibility
Allows for washing of both 96-and 384-well plates. Also includes automated internal buffer switching from up to 4 different buffer bottles.
Combine With SMC® Plate Washer Evaluation Kit For Extra Confidence (Prior to Running Assays)
Utilize our unique plate washer evaluation kit to ensure optimal performance of your washer before running any precious samples.
Learn how to maintain this plate washer in our BioTek® plate washer maintenance article.
Related Products | |
---|---|
Product Description | Product Number |
BioTek® 405™TSUVS Plate Washer | 95-0004-06 |
SMC® Plate Washer Evaluation Kit | 03-0165-00 |
Flat Magnet for 96-Well Plates, for use with MILLIPLEX® Assays | 7103016 |
Flat Magnet for 384-Well Plates, for use with MILLIPLEX® Assays | 7103017 |
For Research Use Only. Not For Use In Diagnostic Procedures.