Merck
UHPLC Analysis of Mesoridazine and Haloperidol on Titan C18
UHPLC Analysis of Mesoridazine and Haloperidol on Titan C18 application for UHPLC
Materials
standard
Product No.
Description
Pricing
Haloperidol
powder
CONDITIONS
column
Titan C18, 10 cm x 2.1 mm I.D., 1.9 μm particles (577124-U)
mobile phase
[A] 10 mM ammonium acetate, pH 7.0; [B] acetonitrile (70:30, A;B)
flow rate
0.3 mL/min
column temp.
35 °C
detector
MS, XIC m/z 387.16 and 376.15
injection
2 μL
sample
300 ng/mL each in acetonitrile:water (3:1)
Description
Legal Information
null