Merck
UHPLC Analysis of B Vitamins in Vitamin Water on Titan C18 using UV Detection
UHPLC Analysis of B Vitamins in Vitamin Water on Titan C18 using UV Detection application for UHPLC
Materials
standard
Product No.
Description
Pricing
Pyridoxine hydrochloride
meets USP testing specifications
Calcium pantothenate
meets USP testing specifications, monograph mol wt. 476.53 (C18H32CaN2O10)
Niacinamide
meets USP testing specifications
Cyanocobalamin (B12)
analytical standard
CONDITIONS
column
TITAN C18, 50 mm x 2.1 mm I.D., 1.9 μm particles (577122-U)
mobile phase
[A] 20 mM potassium phosphate, pH 3; [B] methanol
gradient
0.5% B held for 0.5 minutes, to 30% B in 1.3 minutes, held at 30% B for 1 minute, held at 0.5% B for 0.8 minutes
flow rate
0.5 mL/min
pressure
7977 psi (550 bar)
column temp.
30 °C
detector
UV, 210 nm
injection
0.5 μL
sample
vitamin water was filtered through 0.2um nylon syringe filter
Description
Analysis Note
The vitamin water drink was filtered using 0.2 um Nylon filter and injected into UHPLC for B-vitamin analysis
Legal Information
null