HPLC Analysis of Peptide Antibiotics on Discovery® C8
CONDITIONS
column
Discovery C8, 15 cm × 4.6 mm I.D., 5 μm particles (59353)
mobile phase
[A] 25mM potassium phosphate, pH 3; [B] acetonitrile, (60:40, A:B)
flow rate
1 mL/min
column temp.
35 °C
detector
UV, 220 nm
injection
10 μL 10 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany