Merck
HPLC Analysis of Indapamide Enantiomers on Astec® Cellulose DMP, LC/MS
HPLC Analysis of Indapamide Enantiomers on Astec® Cellulose DMP, LC/MS application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® Cellulose DMP Chiral HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
Indapamide
analytical standard, for drug analysis
CONDITIONS
column
Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase
[A] methanol; [B] ammonium formate; (100:0.1, A:B)
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
2 μl/h
sample
2 mg/mL in mobile phase