Merck
HPLC Analysis of Glyocoalkaloids Solanine and Cacoline on Chromolith® NH2
HPLC Analysis of Glyocoalkaloids Solanine and Cacoline on Chromolith® NH2 application for HPLC
Materials
standard
Product No.
Description
Pricing
α-Solanine
from potato sprouts, ≥95%
CONDITIONS
column
Chromolith NH2, 5 cm x 4.6 mm I.D. (1.52027.0001)
mobile phase
[A] acetonitrile; [B] phosphate buffer; pH 6 (80:20, A:B)
flow rate
2 mL/min
column temp.
23 °C
detector
UV, 208 nm
injection
8 μL
sample
solanine 0.2 mg/mL, cacoline 0.2 mg/mL, in mobile phase
pressure
174 psi (12 bar)
Description
Legal Information
Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany