Merck
HPLC Analysis of Bisphenol A in Spiked Milk Powder on Chromolith® High Resolution RP-18e
HPLC Analysis of Bisphenol A in Spiked Milk Powder on Chromolith® High Resolution RP-18e application for HPLC
CONDITIONS
sample preparation
12.1 g of milik powder dissolved in 100 mL hot water. Spike100 ppb of BPA to the solution, Add 0.1% of HCl and 10% acetonitrile. Add Internal standard BPF 60 ppb. Centrifuged and filter the sample with Mellix simplecity filiter.
column
Chromolith® High Resolution RP-18 encapped, 100 x 4.6 mm (1.52022.0001)
mobile phase
[A] acetonitrile [B] MilliQ water; (25:75, A:B)
flow rate
Flow rate gradient
pressure
1305 psi (90 bar)
column temp.
35 °C
detector
Hitachi Chromaster-5440 Fluorescence Detector, λex=226; λem=306 nm (cell: 13 μL)
injection
10 μL
sample
Bisphenol A & 60ppb Bisphenol F in Methanol(5.2mg/10ml)
Description
Legal Information
Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany