GC Analysis of Fuel Oil #1 on Equity-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
carrier gas
hydrogen, 45 cm/sec constant
column
Equity-1, 15 m x 0.10 mm I.D., 0.10 μm (28039-U)
detector
FID, 350 °C
inj. temp.
250 °C
injection
0.1 μL, 300:1 split, 0.02 min pre-injection dwell time
liner
2 mm I.D., straight
oven
80 °C, 50 °C/min to 325 °C
sample
No.1 Fuel Oil standard, 20 mg/mL in methanol (47518-U)
Description
Legal Information
null