Merck
GC Analysis of a 38-Component Fatty Acid Methyl Ester (FAME) Mix on SLB®-IL60, Comparison to a Polyethylene Glycol (PEG) Column
GC Analysis of a 38-Component Fatty Acid Methyl Ester (FAME) Mix on SLB®-IL60, Comparison to a Polyethylene Glycol (PEG) Column suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-IL60 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
standard
Product No.
Description
Pricing
Methyl all-cis-7,10,13,16,19-docosapentaenoate
analytical standard
F.A.M.E. Mix, C4-C24
certified reference material,  wt. % (varied)
Supelco 37 Component FAME Mix
certified reference material, TraceCERT®, in dichloromethane (varied conc.), ampule of 1 mL
CONDITIONS
column
PEG 3, 30 m x 0.25 mm I.D., 0.25 μm
column
SLB-IL60, 30 m x 0.25 mm I.D., 0.20 μm (29505-U)
oven
170 °C, 1 °C/min to 225 °C
inj. temp.
250 °C
detector
FID, 260 °C
carrier gas
helium, 1.2 mL/min
liner
1.0 μL 100:1 split
liner
4 mm I.D., split/splitless type, single taper wool packed FocusLiner design
sample
Supelco 37-Component FAME Mix (47885-U) + C22:5n3, in methylene chloride
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany