GC Analysis of BTEX, Undecane, and Tridecane on SLB®-IL100
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL100, 30 m × 0.25 mm I.D., 0.20 μm (28884-U)
oven
110 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 26 cm/sec
injection
0.1 μL, 300:1 split
liner
4 mm I.D., split, cup design
sample
neat mixture containing varying percentages of each component
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany