Merck
  • Effects of ultrasound treatment on physico-chemical, functional properties and antioxidant activity of whey protein isolate in the presence of calcium lactate.
Effects of ultrasound treatment on physico-chemical, functional properties and antioxidant activity of whey protein isolate in the presence of calcium lactate.
Journal of the science of food and agriculture (2017-08-13)
Zhanmei Jiang, Kun Yao, Xiangying Yuan, Zhishen Mu, Zengli Gao, Juncai Hou, Lianzhou Jiang
ABSTRACT
The aim of this study was to investigate the effects of ultrasound applied at various powers (0, 200, 400, 600 or 800 W) and for different times (20 or 40 min) on the physico-chemical, functional properties and antioxidant activities of whey protein isolate (WPI) dispersions in the presence of 1.20 mmol L Surface hydrophobicity and free sulfhydryl group of the WPI dispersions containing 1.2 mmol L Ultrasound treatments induced structural changes in WPI molecules, leading to different microstructure and improved gel strength of WPI in the presence of calcium lactate. © 2017 Society of Chemical Industry.
Sigma-Aldrich
1-Naphthalenesulfonic acid, >50%