Merck
Suppressive Effects of
Frontiers in pharmacology (2018-02-17)
Ochuko L Erukainure, Rahman M Hafizur, Nurul Kabir, M Iqbal Choudhary, Olubunmi Atolani, Priyanka Banerjee, Robert Preissner, Chika I Chukwuma, Aliyu Muhammad, Eric O Amonsou, Md Shahidul Islam
ABSTRACT
Type 2 diabetes is the most prominent of all diabetes types, contributing to global morbidity and mortality. Availability and cost of treatment with little or no side effect especially in developing countries, remains a huge burden. This has led to the search of affordable alternative therapies especially from medicinal plants. In this study, the antidiabetic effect of the methanolic extract, dichloromethane (DCM), butanol (BuOH) and aqueous fractions of
Sigma-Aldrich
Cerium(IV) sulfate