Merck
  • ADAM9 is over-expressed in human ovarian clear cell carcinomas and suppresses cisplatin-induced cell death.
ADAM9 is over-expressed in human ovarian clear cell carcinomas and suppresses cisplatin-induced cell death.
Cancer science (2017-12-17)
Mari Ueno, Takayuki Shiomi, Satsuki Mochizuki, Miyuki Chijiiwa, Masayuki Shimoda, Yae Kanai, Fumio Kataoka, Akira Hirasawa, Nobuyuki Susumu, Daisuke Aoki, Yasunori Okada
ABSTRACT
ADAMs (a disintegrin and metalloproteinases) are involved in various biological events such as cell adhesion, migration and invasion, membrane protein shedding and proteolysis. However, there have been no systematic studies on the expression of ADAMs in human ovarian carcinomas. We therefore examined mRNA expression of all the proteolytic ADAM species including ADAM8, 9, 10, 12, 15, 17, 19, 20, 21, 28, 30, 33 and ADAMDEC1 in human ovarian carcinomas, and found that prototype membrane-anchored ADAM9m, but not secreted isoform ADAM9s, is significantly over-expressed in carcinomas than in control non-neoplastic ovarian tissue. Among the histological subtypes of serous, endometrioid, mucinous and clear cell carcinomas, ADAM9m expression was highest in clear cell carcinomas. Immunohistochemistry showed that all the clear cell carcinoma samples displayed ADAM9m primarily on the carcinoma cell membrane. By immunoblotting, ADAM9m was detected mainly in an active form in the clear cell carcinoma tissues. When two clear cell carcinoma cell lines (RMG-I and TOV21G cells) with ADAM9m expression were treated with cisplatin, viability was significantly reduced and apoptosis increased in ADAM9m knockdown cells compared with mock transfectants. In addition, treatment of the cells with neutralizing anti-ADAM9m antibody significantly decreased viability compared with non-immune IgG, whereas ADAM9m over-expression significantly increased viability compared with mock transfectants. Our data show, to the best of our knowledge, for the first time, that ADAM9m is over-expressed in an activated form in human ovarian clear cell carcinomas, and suggest that ADAM9m plays a key role in cisplatin resistance.
Sigma-Aldrich
MISSION® esiRNA, targeting human ADAM9
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid