- Lymphangiogenesis in rat asthma model.
Lymphangiogenesis in rat asthma model.
Angiogenesis (2016-10-28)
Aigul Moldobaeva, John Jenkins, Qiong Zhong, Elizabeth M Wagner
PMID27787629
ABSTRACT
Although bronchial angiogenesis has been well documented in allergic asthma, lymphangiogenesis has not been widely studied. Therefore, we evaluated changes in lung lymphatics in a rat model of allergen-induced asthma using house dust mite (Der p 1; 100 μg/challenge). Additionally, properties of isolated lung lymphatic endothelial cells (CD45
Sigma-Aldrich
Anti-Potassium Channel Kv11.1 (HERG) Extracellular−FITC antibody produced in rabbit, affinity isolated antibody, lyophilized powder