Merck
  • The PRMT5/WDR77 complex regulates alternative splicing through ZNF326 in breast cancer.
The PRMT5/WDR77 complex regulates alternative splicing through ZNF326 in breast cancer.
Nucleic acids research (2017-10-05)
Madhumitha Rengasamy, Fan Zhang, Ajay Vashisht, Won-Min Song, Francesca Aguilo, Yifei Sun, SiDe Li, Weijia Zhang, Bin Zhang, James A Wohlschlegel, Martin J Walsh
ABSTRACT
We observed overexpression and increased intra-nuclear accumulation of the PRMT5/WDR77 in breast cancer cell lines relative to immortalized breast epithelial cells. Utilizing mass spectrometry and biochemistry approaches we identified the Zn-finger protein ZNF326, as a novel interaction partner and substrate of the nuclear PRMT5/WDR77 complex. ZNF326 is symmetrically dimethylated at arginine 175 (R175) and this modification is lost in a PRMT5 and WDR77-dependent manner. Loss of PRMT5 or WDR77 in MDA-MB-231 cells leads to defects in alternative splicing, including inclusion of A-T rich exons in target genes, a phenomenon that has previously been observed upon loss of ZNF326. We observed that the alternatively spliced transcripts of a subset of these genes, involved in proliferation and tumor cell migration like REPIN1/AP4, ST3GAL6, TRNAU1AP and PFKM are degraded upon loss of PRMT5. In summary, we have identified a novel mechanism through which the PRMT5/WDR77 complex maintains the balance between splicing and mRNA stability through methylation of ZNF326.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-dimethyl-Arginine Antibody, symmetric (SYM10), serum, Upstate®