Merck
  • TFP5 prevents 1-methyl-4-phenyl pyridine ion-induced neurotoxicity in mouse cortical neurons.
TFP5 prevents 1-methyl-4-phenyl pyridine ion-induced neurotoxicity in mouse cortical neurons.
Experimental and therapeutic medicine (2016-10-05)
Qi-Shan Zhang, Yuan-Gao Liao, Zhong Ji, Yong Gu, Hai-Shan Jiang, Zuo-Shan Xie, Su-Yue Pan, Ya-Fang Hu
ABSTRACT
The present study aimed to investigate the protective effect of a modified p5 peptide, TFP5, on 1-methyl-4-phenyl pyridine ion (MPP
Sigma-Aldrich
Anti-phospho-MEF2D (pSer444) antibody produced in rabbit, affinity isolated antibody