Merck
  • Exploration of the expeditious potential of Pseudomonas fluorescens lipase in the kinetic resolution of racemic intermediates and its validation through molecular docking.
Exploration of the expeditious potential of Pseudomonas fluorescens lipase in the kinetic resolution of racemic intermediates and its validation through molecular docking.
Chirality (2017-10-25)
Surbhi Soni, Bharat P Dwivedee, Vishnu K Sharma, Gopal Patel, Uttam C Banerjee
ABSTRACT
A profoundly time-efficient chemoenzymatic method for the synthesis of (S)-3-(4-chlorophenoxy)propan-1,2-diol and (S)-1-chloro-3-(2,5-dichlorophenoxy)propan-2-ol, two important pharmaceutical intermediates, was successfully developed using Pseudomonas fluorescens lipase (PFL). Kinetic resolution was successfully achieved using vinyl acetate as acylating agent, toluene/hexane as solvent, and reaction temperature of 30Ā°C giving high enantioselectivity and conversion. Under optimized condition, PFL demonstrated 50.2% conversion, enantiomeric excess of 95.0%, enantioselectivity (EĀ =Ā 153) in an optimum time of 1Ā hour and 50.3% conversion, enantiomeric excess of 95.2%, enantioselectivity (EĀ =Ā 161) in an optimum time of 3Ā hours, for the two racemic alcohols, respectively. Docking of the R- and S-enantiomers of the intermediates demonstrated stronger H-bond interaction between the hydroxyl group of the R-enantiomer and the key binding residues of the catalytic site of the lipase, while the S-enantiomer demonstrated lesser interaction. Thus, docking study complemented the experimental outcome that PFL preferentially acylated the R form of the intermediates. The present study demonstrates a cost-effective and expeditious biocatalytic process that can be applied in the enantiopure synthesis of pharmaceutical intermediates and drugs.
Sigma-Aldrich
Cyclopropanemethanol, ≥99.5%