Merck
Farnesyl phenolic enantiomers as natural MTH1 inhibitors from
Oncotarget (2017-12-10)
Ya Gao, Lihan Zhu, Jing Guo, Ting Yuan, Liqing Wang, Hua Li, Lixia Chen
ABSTRACT
Cancer cells are more addictive to MTH1 than normal cells because of their dysfunctional redox regulations. MTH1 plays an important role to maintain tumor cell survival, while it is not indispensable for the growth of normal cells. Farnesyl phenols having a coumaroyl substitution are rather uncommon in nature. Eight farnesyl phenolic compounds with such substituent moiety (1-8), including six new ones, ganosinensols E-J (1-6) were isolated from the 95% EtOH extract of the fruiting bodies of
Sigma-Aldrich
MISSION® esiRNA, targeting human NUDT1