Merck
  • A green ionic liquid-based vortex-forced MSPD method for the simultaneous determination of 5-HMF and iridoid glycosides from Fructus Corni by ultra-high performance liquid chromatography.
A green ionic liquid-based vortex-forced MSPD method for the simultaneous determination of 5-HMF and iridoid glycosides from Fructus Corni by ultra-high performance liquid chromatography.
Food chemistry (2017-11-10)
Kunze Du, Jin Li, Yun Bai, Mingrui An, Xiu-Mei Gao, Yan-Xu Chang
ABSTRACT
A simple and green ionic liquid-based vortex-forced matrix solid phase dispersion (IL-VFMSPD) method was presented to simultaneously extract 5-hydroxymethyl furfurol (5-HMF) and iridoid glycosides in Fructus Corni by ultra-high performance liquid chromatography. Ionic liquid was used as a green elution reagent in vortex-forced MSPD process. A few parameters such as the type of ionic liquid, the type of sorbent, ratio of sample to sorbent, the concentration and volume of ionic liquid, grinding time and vortex time, were investigated in detail and an orthogonal design experiment was introduced to confirm the best conditions in this procedure. With the final optimized method, the recoveries of the target compounds in Fructus Corni were in the range of 95.2-103% (RSD<5.0%) and the method displayed a good linearity within the range of 0.8-200 μg mL
Sigma-Aldrich
1-Hexyl-3-methylimidazolium tetrafluoroborate, ≥97.0% (HPLC)
Sigma-Aldrich
1-Hexyl-3-methylimidazolium hexafluorophosphate, ≥97.0% (HPLC)
Sigma-Aldrich
1-Methyl-3-octylimidazolium hexafluorophosphate, ≥95.0%
Sigma-Aldrich
1-Methyl-3-octylimidazolium tetrafluoroborate, ≥97.0% (HPLC)