- TAK1 inhibition attenuates both inflammation and fibrosis in experimental pneumoconiosis.
TAK1 inhibition attenuates both inflammation and fibrosis in experimental pneumoconiosis.
Pneumoconiosis, caused by inhalation of mineral dusts, is a major occupational disease worldwide. Currently, there are no effective drugs owing to a lack of potential therapeutic targets during either the inflammation or fibrosis molecular events in pneumoconiosis. Here, we performed microarrays to identify aberrantly expressed genes in the above molecular events