Merck
  • CR6-interacting factor 1 inhibits invasiveness by suppressing TGF-β-mediated epithelial-mesenchymal transition in hepatocellular carcinoma.
CR6-interacting factor 1 inhibits invasiveness by suppressing TGF-β-mediated epithelial-mesenchymal transition in hepatocellular carcinoma.
Oncotarget (2017-12-08)
Runzhou Zhuang, Di Lu, Jianyong Zhuo, Xuanyu Zhang, Kun Wang, Xuyong Wei, Qiang Wei, Wei Wang, Haiyang Xie, Lin Zhou, Xiao Xu, Shusen Zheng
ABSTRACT
CR6-interacting factor 1 (CRIF1) regulates cell cycle progression and the DNA damage response. Here, we show that CRIF1 expression is decreased in hepatocellular carcinoma (HCC) tissues and positively correlates with patients' survival.
Sigma-Aldrich
MISSION® esiRNA, targeting human CHN1
Sigma-Aldrich
MISSION® esiRNA, targeting human GADD45GIP1
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid