Merck
  • An experimentally validated network of nine haematopoietic transcription factors reveals mechanisms of cell state stability.
An experimentally validated network of nine haematopoietic transcription factors reveals mechanisms of cell state stability.
eLife (2016-02-24)
Judith Schütte, Huange Wang, Stella Antoniou, Andrew Jarratt, Nicola K Wilson, Joey Riepsaame, Fernando J Calero-Nieto, Victoria Moignard, Silvia Basilico, Sarah J Kinston, Rebecca L Hannah, Mun Chiang Chan, Sylvia T Nürnberg, Willem H Ouwehand, Nicola Bonzanni, Marella Ftr de Bruijn, Berthold Göttgens
ABSTRACT
Transcription factor (TF) networks determine cell-type identity by establishing and maintaining lineage-specific expression profiles, yet reconstruction of mammalian regulatory network models has been hampered by a lack of comprehensive functional validation of regulatory interactions. Here, we report comprehensive ChIP-Seq, transgenic and reporter gene experimental data that have allowed us to construct an experimentally validated regulatory network model for haematopoietic stem/progenitor cells (HSPCs). Model simulation coupled with subsequent experimental validation using single cell expression profiling revealed potential mechanisms for cell state stabilisation, and also how a leukaemogenic TF fusion protein perturbs key HSPC regulators. The approach presented here should help to improve our understanding of both normal physiological and disease processes.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tal1