Merck
  • Photosystem II-cyclic electron flow powers exceptional photoprotection and record growth in the microalga Chlorella ohadii.
Photosystem II-cyclic electron flow powers exceptional photoprotection and record growth in the microalga Chlorella ohadii.
Biochimica et biophysica acta (2017-07-25)
Gennady Ananyev, Colin Gates, Aaron Kaplan, G Charles Dismukes
ABSTRACT
The desert microalga Chlorella ohadii was reported to grow at extreme light intensities with minimal photoinhibition, tolerate frequent de/re-hydrations, yet minimally employs antenna-based non-photochemical quenching for photoprotection. Here we investigate the molecular mechanisms by measuring Photosystem II charge separation yield (chlorophyll variable fluorescence, Fv/Fm) and flash-induced O
Sigma-Aldrich
2,6-Dimethoxy-1,4-benzoquinone, 97%