Merck
  • Inactivation of viruses and bacteria on strawberries using a levulinic acid plus sodium dodecyl sulfate based sanitizer, taking sensorial and chemical food safety aspects into account.
Inactivation of viruses and bacteria on strawberries using a levulinic acid plus sodium dodecyl sulfate based sanitizer, taking sensorial and chemical food safety aspects into account.
International journal of food microbiology (2017-07-03)
Zijin Zhou, Sophie Zuber, Frédérique Cantergiani, Sophie Butot, Dan Li, Thomas Stroheker, Frank Devlieghere, Anthony Lima, Umberto Piantini, Mieke Uyttendaele
ABSTRACT
The efficacy of levulinic acid (LVA) in combination with sodium dodecyl sulfate (SDS) in removal of foodborne viruses, enteric bacterial pathogens and their surrogates on fresh strawberries was investigated. Inoculated strawberries were treated with potable water, sodium hypochlorite solution (50ppm), 0.5% LVA plus 0.5% SDS solution, and 5% LVA plus 2% SDS solution respectively for 2min, followed by spray-rinsing with potable water. Water washing removed at least 1.0-log of the tested viral and bacterial strains from the strawberries' surfaces. The 50ppm chlorine wash induced 3.4, 1.5 and 2.1-log reductions for hepatitis A virus (HAV), murine norovirus-1 (MNV-1) and MS2 bacteriophage, respectively. In comparison, the tested bacterial strains showed uniform reductions around 1.6-log CFU/ml. The 0.5% LVA plus 0.5% SDS wash induced 2.7, 1.4 and 2.4-log reductions for HAV, MNV-1 and MS2, which were comparable with the reductions induced by chlorine (P>0.05). For bacteria, over 2.0-log reductions were obtained for Enterococcus faecium, Listeria monocytogenes and Salmonella, while Escherichia coli O157:H7 and Escherichia coli P1 showed reductions of 1.9 and 1.8-log CFU/ml. Higher concentration of LVA plus SDS showed no significantly higher reductions (P>0.05). Sensory tests of washed strawberries and chemical residue analysis of LVA on strawberries after washing were also performed. In conclusion, this study demonstrates good performance of 0.5% LVA plus 0.5% SDS to reduce the levels of enteric pathogens if present on strawberries without altering taste and introducing chemical safety issues.
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Levulinic acid, 98%