Merck
  • Silica- and germania-based dual-ligand sol-gel organic-inorganic hybrid sorbents combining superhydrophobicity and π-π interaction. The role of inorganic substrate in sol-gel capillary microextraction.
Silica- and germania-based dual-ligand sol-gel organic-inorganic hybrid sorbents combining superhydrophobicity and π-π interaction. The role of inorganic substrate in sol-gel capillary microextraction.
Analytica chimica acta (2017-03-30)
Emre Seyyal, Abdul Malik
ABSTRACT
Principles of sol-gel chemistry were utilized to create silica- and germania-based dual-ligand surface-bonded sol-gel coatings providing enhanced performance in capillary microextraction (CME) through a combination of ligand superhydrophobicity and π-π interaction. These organic-inorganic hybrid coatings were prepared using sol-gel precursors with bonded perfluorododecyl (PF-C
Sigma-Aldrich
Butylcyclohexane, ≥99%
Sigma-Aldrich
Propylbenzene, 98%
Sigma-Aldrich
Trimethoxy(2-phenylethyl)silane, 98%