Merck
  • Triphenylphosphonium-modified poly(ethylene glycol)-poly(ε-caprolactone) micelles for mitochondria- targeted gambogic acid delivery.
Triphenylphosphonium-modified poly(ethylene glycol)-poly(ε-caprolactone) micelles for mitochondria- targeted gambogic acid delivery.
International journal of pharmaceutics (2017-02-22)
Yingqi Xu, Shengpeng Wang, Hon Fai Chan, Yuling Liu, Hui Li, Chengwei He, Zeyong Li, Meiwan Chen
ABSTRACT
Mitochondria are important targets for the intracellular delivery of drugs and DNA. For mitochondria-targeted delivery, a mitochondriotropic molecule, triphenylphosphonium (TPP), was applied to the synthesis of amphiphilic TPP-poly(ethylene glycol)-poly(ε-caprolactone) (TPP-PEG-PCL) polymers. The TPP-PEG-PCL polymer was used to prepare micelles using a solvent evaporation method for the delivery of gambogic acid (GA) (GA-TPP). The micelles were obtained with a favorable particle size of 150.07±11.71nm and an encapsulation efficiency of 80.78±1.36%, and they displayed homogeneous spherical shapes. The GA-TPP micelles exerted enhanced cytotoxic and pro-apoptotic effect against A549 cells compared to free GA and GA-loaded PEG-PCL (GA-PP) micelles, due to the inhibition of the expression of apoptosis-related proteins and promotion of caspase 3/7 and caspase 9 activity. Notably, the mitochondria-targeting GA-TPP micelles selectively accumulated in the mitochondria, inducing the loss of mitochondrial membrane potential and the release of cytochrome c, thereby achieving improved mitochondria-targeting effects. In conclusion, the GA-TPP micelle system shows great promise for lung cancer treatment by inducing an apoptotic effect via the mitochondrial signaling pathway.
Sigma-Aldrich
2-(Dimethylamino)pyridine, 97%
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)