Merck
  • Determination of volatile organic compounds in pen inks by a dynamic headspace needle trap device combined with gas chromatography-mass spectrometry.
Determination of volatile organic compounds in pen inks by a dynamic headspace needle trap device combined with gas chromatography-mass spectrometry.
Journal of chromatography. A (2017-07-25)
Xiaohuan Zang, Weiqian Liang, Qingyun Chang, Tong Wu, Chun Wang, Zhi Wang
ABSTRACT
Some harmful volatile organic compounds (VOCs), such as methylbenzene, ethylbenzene, xylene, chlorobenzene and bromobenzene, are the commonly found chemicals in pen inks. In this work, a dynamic headspace needle-trap device (D-HS-NTD) with a ZIF-8-derived nanoporous carbon (ZIF-8-NPC) as the adsorbent was developed for the extraction of some VOCs in different pen inks prior to GC-MS detection. The main important influencing experimental parameters including the flow rate of the purge gas N
Sigma-Aldrich
Triethanolamine oleate, AldrichCPR
Sigma-Aldrich
D.E.R. 332, used as embedding medium